Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

2,5-Dichloropyridine-3-carbonyl chloride, 97%, Thermo Scientific™

Catalog No. SPB06842EA
Change view
Click to view available options
Quantity:
10 g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
SPB06842EA 10 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. SPB06842EA Supplier Thermo Scientific Chemicals Supplier No. SPB06842EA
Add to Cart

Chemical Identifiers

CAS 78686-87-0
Molecular Formula C6H2Cl3NO
Molecular Weight (g/mol) 210.438
MDL Number MFCD00210149
InChI Key VRJMAKCJTSZUQR-UHFFFAOYSA-N
Synonym 2,5-dichloronicotinoyl chloride, 2,5-dichloro-nicotinoyl chloride, 2,5-dichloro-3-pyridinecarbonyl chloride, 2,5-dichloronicotinic acid chloride, 2,5-dichloropyridine-3-carbonylchloride, 3-chlorocarbonyl-2,5-dichloropyridine, 2,5-dichloro-pyridine-3-carbonyl chloride, 3-pyridinecarbonylchloride, 2,5-dichloro, 3-pyridinecarbonyl chloride, 2,5-dichloro
PubChem CID 2799996
IUPAC Name 2,5-dichloropyridine-3-carbonyl chloride
SMILES C1=C(C(=NC=C1Cl)Cl)C(=O)Cl

Specifications

CAS Min % 97.0
Assay Percent Range 97%
Packaging Amber glass bottle
Quantity 10 g
Formula Weight 210.45
Percent Purity 97%
Chemical Name or Material 2, 5-Dichloropyridine-3-carbonyl chloride
Hazard Category GHS Signal Word: Danger
Hazard Statement GHS H Statement:
Causes severe skin burns and eye damage.
Causes serious eye damage.
Precautionary Statement GHS P Statement:
IF SWALLOWED: rinse mouth. Do NOT induce vomiting.
Wear eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
Immediately call a POISON CENTER or doctor/physician.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.