Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

2,5-Dichloropyridine, 98%, ACROS Organics™

Click to view available options
Quantity:
5g
25g
100g
Packaging:
Glass bottle
16110-09-1
C5H3Cl2N
147.99
MFCD00006239
GCTFDMFLLBCLPF-UHFFFAOYSA-N
pyridine, 2,5-dichloro,2,5-dichloro-pyridine,2,5-dichloro pyridine,ccris 1718,2,5-dichlorpyridin,2,5-dichloropyridin,2,5-dichioropyridine,2.5-dichloropyridine,pubchem1199,2,5 dichloropyridine
27685
2,5-dichloropyridine
C1=CC(=NC=C1Cl)Cl
16110-09-1
C5H3Cl2N
147.99
MFCD00006239
GCTFDMFLLBCLPF-UHFFFAOYSA-N
pyridine, 2,5-dichloro,2,5-dichloro-pyridine,2,5-dichloro pyridine,ccris 1718,2,5-dichlorpyridin,2,5-dichloropyridin,2,5-dichioropyridine,2.5-dichloropyridine,pubchem1199,2,5 dichloropyridine
27685
2,5-dichloropyridine
C1=CC(=NC=C1Cl)Cl
Packaging Glass bottle
Water 1% max.
Flash Point 112°C
Infrared Spectrum Authentic
Quantity 100g
Density 112g/mL
Boiling Point 190.0°C to 191.0°C
Melting Point 58.0°C to 62.0°C
Color White to Yellow
CAS Min % 97.5
CAS Max % 100.0
Assay Percent Range 97.5% min. (GC)
Linear Formula (C5H3N)Cl2
Beilstein 20, I, 80
Solubility Information Solubility in water: insoluble
Formula Weight 147.99
Physical Form Crystalline Solid
Percent Purity 98%
Chemical Name or Material 2, 5-Dichloropyridine
Show More Show Less

GHS H Statement:
Harmful if swallowed.
May cause respiratory irritation.
Causes skin irritation.
Causes serious eye irritation.

GHS P Statement:
Avoid breathing dust/fume/gas/mist/vapors/spray.
IF SWALLOWED: Call a POISON CENTER or doctor/physician if you feel unwell.
IF ON SKIN: Wash with plenty of soap and water.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.

GHS Signal Word: Warning

EINECSNumber : 240-278-2