Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

2,5-Dichlorothiophene, 98%, Thermo Scientific™

Catalog No. AAA1383936
Change view
Click to view available options
Quantity:
500g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AAA1383936 500g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. AAA1383936 Supplier Thermo Scientific Chemicals Supplier No. A1383936

missing translation for 'validMainframeMsg'

This Thermo Scientific brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.

TRUSTED_SUSTAINABILITY

Specifications

CAS 3172-52-9
Density 1.442
Boiling Point 160°C to 162°C
Flash Point 59°C (138°F)
Odor Pungent
Molecular Formula C4H2Cl2S
Refractive Index 1.562
Quantity 500g
MDL Number MFCD00005423
UN Number UN2929
Beilstein 109887
Synonym thiophene, 2,5-dichloro, 2,-dichlorothiophene, 2,5-dichloro-thiophene, pubchem5521, thiophene,5-dichloro, 2,5-dichloro thiophene, thiophene,2,5-dichloro, acmc-1cmg3, ksc223o3n, bidd:gt0088
InChI Key FGYBDASKYMSNCX-UHFFFAOYSA-N
SMILES C1=C(SC(=C1)Cl)Cl
IUPAC Name 2,5-dichlorothiophene
Molecular Weight (g/mol) 153.02
PubChem CID 18501
Formula Weight 153.03
Percent Purity 98%
Chemical Name or Material 2,5-Dichlorothiophene
Show More Show Less
Hazard Category H226-H300+H310+H330-H351
Hazard Statement GHS H Statement
H300-H310-H226-H341
Fatal if swallowed.
Fatal in contact with skin.
Flammable liquid and vapour.
Suspected of causing genetic defects.
Precautionary Statement P201-P202-P210-P233-P235-P240-P241-P242-P243-P260-P262-P264b-P270-P271-P281-P284-P301+P310-P303+P361+P353-P304+P340-P308+P313-P310-P330-P363-P370+P378q-P501c
DOTInformation Transport Hazard Class: 6.1; Packing Group: II; Proper Shipping Name: TOXIC LIQUIDS, FLAMMABLE, ORGANIC, N.O.S.
EINECSNumber 221-638-8
TSCA Yes
Recommended Storage Keep cold
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.