Learn More
Cayman Chemical 2 5-dIdeoxy AdenosIn 10mg
SDP
Catalog No. 502984090
Encompass_Preferred
A cell-permeable, P-site inhibitor of adenylate cyclase; inhibits forskolin-induced activation of a cAMP-dependent reporter gene in HEK293 cells with an IC50 value of 33 µM
Catalog No. 50-298-4090 Supplier Cayman Chemical Supplier No. 2035810 MG
A cell-permeable, P-site inhibitor of adenylate cyclase; inhibits forskolin-induced activation of a cAMP-dependent reporter gene in HEK293 cells with an IC50 value of 33 µM