Learn More
Apexbio Technology LLC 2-5-DIDEOXY-ADENOSINE-5MG
SDP
Catalog No. 5000568650
Encompass_Preferred
2 5 -dideoxy Adenosine (CAS No 6698-26-6) is a nucleoside analogue commonly studied in the field of biochemical research due to its unique properties affecting adenosine pathways Originating from modifications of adenosine it disrupts various cellular mechanisms by targeting adenosine receptors and influencing intracellular signaling Its activity is typically observed in the nanomolar to low micromolar range making it relevant for analyzing cellular responses and biochemical processes Frequently utilized in vitro it serves as a tool in various cell models including human cell lines and its influence is often assessed in drug screening assays to understand its potential therapeutic implications Studies also extend to animal models to evaluate its biological effects and pharmacokinetics Experimental concentrations typically depend on the specific research design ensuring adaptability across diverse scientific investigations
Catalog No. 50-005-68650 Supplier Apexbio Technology LLC Supplier No. C39935
2 5 -dideoxy Adenosine (CAS No 6698-26-6) is a nucleoside analogue commonly studied in the field of biochemical research due to its unique properties affecting adenosine pathways Originating from modifications of adenosine it disrupts various cellular mechanisms by targeting adenosine receptors and influencing intracellular signaling Its activity is typically observed in the nanomolar to low micromolar range making it relevant for analyzing cellular responses and biochemical processes Frequently utilized in vitro it serves as a tool in various cell models including human cell lines and its influence is often assessed in drug screening assays to understand its potential therapeutic implications Studies also extend to animal models to evaluate its biological effects and pharmacokinetics Experimental concentrations typically depend on the specific research design ensuring adaptability across diverse scientific investigations