Learn More
TARGETMOL CHEMICALS INC 2 5 -Dideoxyadenosine 10MG
SDP
Catalog No. 502880679
Encompass_Preferred
Also available in 1 mL, 1 mg, 5 mg, 25 mg, 50 mg, 100 mg, 500 mg and bulk. Please contact Fisher for quotes. 2',5'-Dideoxyadenosine is a potent and non-competitive adenylyl cyclase inhibitor, binding to the P-site with an IC50 of 3 uM. Purity 99.55%
Catalog No. 50-288-0679 Supplier TARGETMOL CHEMICALS INC Supplier No. T1006610MG
Also available in 1 mL, 1 mg, 5 mg, 25 mg, 50 mg, 100 mg, 500 mg and bulk. Please contact Fisher for quotes. 2',5'-Dideoxyadenosine is a potent and non-competitive adenylyl cyclase inhibitor, binding to the P-site with an IC50 of 3 uM. Purity 99.55%