Learn More
Enzo Life Sciences 2',5'-Dideoxyadenosine (25mg). CAS: 6698-26-6
SDP
Catalog No. 502010439
Encompass_Preferred
A useful, cell-permeable inhibitor of adenylate cyclase (ID50=250µM). Inhibits growth and causes cell differentiation (1.5µM) in K562 cells. Purity: ≥98% (HPLC). Solubility: Soluble in water (20mg/ml at 50°C) or DMSO (30mg/ml). Long Term Storage: -20°C.
Catalog No. 50-201-0439 Supplier Enzo Life Sciences Supplier No. BMLCN1100025
A useful, cell-permeable inhibitor of adenylate cyclase (ID50=250µM). Inhibits growth and causes cell differentiation (1.5µM) in K562 cells. Purity: ≥98% (HPLC). Solubility: Soluble in water (20mg/ml at 50°C) or DMSO (30mg/ml). Long Term Storage: -20°C.