Learn More
Santa Cruz Biotechnology 2′,5′-Dideoxyadenosine, 6698-26-6, MFCD00210904, 5 mg
Catalog No. NC9932632
Encompass
Molecular Weight: 235.2, Molecular Formula: C10H13N5O2, ≥98%, Store at -20° C, MP: 176-178° C, BP: ~546.96° C. A cell-permeable inhibitor of adenylate cyclase, a protein involved in cellular signaling and mast cell granule secretion in immunologic response.
Catalog No. NC9932632 Supplier Santa Cruz Biotechnology Supplier No. SC201562
Molecular Weight: 235.2, Molecular Formula: C10H13N5O2, ≥98%, Store at -20° C, MP: 176-178° C, BP: ~546.96° C. A cell-permeable inhibitor of adenylate cyclase, a protein involved in cellular signaling and mast cell granule secretion in immunologic response.