Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

2,5-Diethoxyaniline 98.0+%, TCI America™
SDP

Catalog No. D045025G Shop All Tokyo Chemical Industry (TCI) Products
Change view
Click to view available options
Quantity:
25 g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
D045025G 25 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. D045025G Supplier TCI America Supplier No. D045025G
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

Chemical Identifiers

CAS 94-85-9
Molecular Formula C10H15NO2
Molecular Weight (g/mol) 181.235
MDL Number MFCD00015144
InChI Key XPKFTIYOZUJAGA-UHFFFAOYSA-N
Synonym benzenamine, 2,5-diethoxy, aminohydroquinone diethyl ether, 2,5-diethoxy aniline, aniline, 2,5-diethoxy, 2,5-diethoxyphenyl amine, 2,5-diethoxyphenylamine, 2.5-diethoxyaniline, aniline,5-diethoxy, 2,5-diathoxy-anilin, 2,5-diethoxybenzenamine
PubChem CID 66756
IUPAC Name 2,5-diethoxyaniline
SMILES CCOC1=CC(=C(C=C1)OCC)N

Specifications

Melting Point 85°C
Color Red-Yellow
Quantity 25 g
Formula Weight 181.24
Percent Purity ≥98.0% (GC,T)
Physical Form Crystalline Powder
Chemical Name or Material 2,5-Diethoxyaniline
EINECSNumber (3)-0616
TSCA Yes
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.