Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

2',5'-Difluoroacetophenone 98.0+%, TCI America™
SDP

Catalog No. D26075G Shop All Tokyo Chemical Industry (TCI) Products
Change view
Click to view available options
Quantity:
5 g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
D26075G 5 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. D26075G Supplier TCI America Supplier No. D26075G
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

Chemical Identifiers

CAS 1979-36-8
Molecular Formula C8H6F2O
Molecular Weight (g/mol) 156.132
MDL Number MFCD00009898
InChI Key HLAFIZUVVWJAKL-UHFFFAOYSA-N
Synonym 2',5'-difluoroacetophenone, 1-2,5-difluorophenyl ethanone, 2,5-difluoroacetophenone, 1-2,5-difluorophenyl ethan-1-one, ethanone, 1-2,5-difluorophenyl, 1-acetyl-2,5-difluorobenzene, fr df bv1, pubchem8394, 2,5-difluoro acetophenone, acmc-209f2b
PubChem CID 74794
IUPAC Name 1-(2,5-difluorophenyl)ethanone
SMILES CC(=O)C1=C(C=CC(=C1)F)F

Specifications

Color Yellow
Boiling Point 61°C
Quantity 5 g
Formula Weight 156.13
Percent Purity ≥98.0% (GC)
Physical Form Liquid
Chemical Name or Material 2′,5′-Difluoroacetophenone
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.