Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

2,5-Difluoroaniline 98.0+%, TCI America™
SDP

Catalog No. D163425G Shop All Tokyo Chemical Industry (TCI) Products
Change view
Click to view available options
Quantity:
5 g
25 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
D163425G 25 g
D16345G 5 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. D163425G Supplier TCI America Supplier No. D163425G
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

Chemical Identifiers

CAS 367-30-6
Molecular Formula C6H5F2N
Molecular Weight (g/mol) 129.11
MDL Number MFCD00007651
InChI Key YNOOQIUSYGWMSS-UHFFFAOYSA-N
Synonym benzenamine, 2,5-difluoro, 2,5-difluorobenzenamine, 2,5-difluoro-phenylamine, aniline, 2,5-difluoro, 2.5-difluoroaniline, aniline,5-difluoro, pubchem1531, 2,5-difluoro-aniline, 2,5-difluorophenylamine, benzenamine,5-difluoro
PubChem CID 67775
IUPAC Name 2,5-difluoroaniline
SMILES C1=CC(=C(C=C1F)N)F

Specifications

Boiling Point 174°C
Quantity 25 g
UN Number 2810
Formula Weight 129.11
Percent Purity ≥98.0% (GC)
Physical Form Liquid
Chemical Name or Material 2,5-Difluoroaniline
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.