Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

2,5-Difluoroanisole 98.0+%, TCI America™
SDP

Catalog No. D380525G Shop All Tokyo Chemical Industry (TCI) Products
Change view
Click to view available options
Quantity:
25 g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
D380525G 25 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. D380525G Supplier TCI America Supplier No. D380525G

Chemical Identifiers

CAS 75626-17-4
Molecular Formula C7H6F2O
Molecular Weight (g/mol) 144.121
MDL Number MFCD00042561
InChI Key HUDMAQLYMUKZOZ-UHFFFAOYSA-N
PubChem CID 2724989
IUPAC Name 1,4-difluoro-2-methoxybenzene
SMILES COC1=C(C=CC(=C1)F)F

Specifications

Color Yellow
Boiling Point 73°C
Quantity 25 g
UN Number 1993
Synonym 2,5-difluoroanisole, 2,5-difluoromethoxybenzene, benzene, 1,4-difluoro-2-methoxy, 2,5-difluoro-1-methoxybenzene, 2,5-difluoroanisol, pubchem3456, acmc-209oz7, ksc641o8p, 1,4-difluoro-2-methoxy-benzene, benzene,1,4-difluoro-2-methoxy
Formula Weight 144.12
Percent Purity ≥98.0% (GC)
Physical Form Liquid
Chemical Name or Material 2,5-Difluoroanisole
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.