Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

2,5-Difluorobenzaldehyde, 98%, Thermo Scientific Chemicals

Catalog No. AC303520010
Change view
Click to view available options
Quantity:
1 g
5 g
25 g
Packaging:
Glass bottle
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity Packaging
AC303520010 1 g Glass bottle
AC303520050 5 g Glass bottle
AC303520250 25 g Glass bottle
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Catalog No. AC303520010 Supplier Thermo Scientific Chemicals Supplier No. 303520010
Get Up to $1,800 in Chemicals with Purchase
View all promotions
Add to Cart

missing translation for 'validMainframeMsg'

CAS: 2646-90-4 | C7H4F2O | 142.11 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 2646-90-4
Molecular Formula C7H4F2O
Molecular Weight (g/mol) 142.11
MDL Number MFCD00010327
InChI Key VVVOJODFBWBNBI-UHFFFAOYSA-N
Synonym 2,5-difluoro benzaldehyde, 2,5-difluoro-benzaldehyde, 2,5-difluorobenzaldehyde, benzaldehyde, 2,5-difluoro, timtec-bb sbb006570, 2,5-difluorobenzalde, pubchem1437, 2,5-fluorobenzaldehyde, 2,5 difluorobenzaldehyde, acmc-1cq4a
PubChem CID 137663
IUPAC Name 2,5-difluorobenzaldehyde
SMILES C1=CC(=C(C=C1F)C=O)F

Specifications

CAS Min % 97.5
CAS Max % 100.0
Color Colorless to Yellow
Density 1.3000g/mL
Boiling Point 67.0°C to 69.0°C (17.0 mmHg)
Flash Point 58°C
Infrared Spectrum Authentic
Assay Percent Range 97.5% min. (GC)
Packaging Glass bottle
Refractive Index 1.4980 to 1.5000
Linear Formula F2C6H3CHO
Quantity 1 g
Specific Gravity 1.3
Formula Weight 142.11
Percent Purity 98%
Physical Form Liquid
Chemical Name or Material 2, 5-Difluorobenzaldehyde
Show More Show Less
Hazard Category GHS Signal Word: Warning
Hazard Statement GHS H Statement:
Causes serious eye irritation.
May cause respiratory irritation.
Causes skin irritation.
Flammable liquid and vapor.
Precautionary Statement GHS P Statement:
Avoid breathing dust/fume/gas/mist/vapors/spray.
IF ON SKIN: Wash with plenty of soap and water.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
Keep away from heat/sparks/open flames/hot surfaces. - No smoking.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.