Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

2,5-Difluorobenzyl Cyanide 98.0+%, TCI America™
SDP

Catalog No. D34425G Shop All Tokyo Chemical Industry (TCI) Products
Change view
Click to view available options
Quantity:
5 g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
D34425G 5 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. D34425G Supplier TCI America Supplier No. D34425G
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

Chemical Identifiers

CAS 69584-87-8
Molecular Formula C8H5F2N
Molecular Weight (g/mol) 153.13
MDL Number MFCD00009972
InChI Key UIMMFRUOZOWROM-UHFFFAOYSA-N
Synonym 2,5-difluorophenylacetonitrile, 2,5-difluorobenzyl cyanide, 2-2,5-difluorophenyl acetonitrile, 2,5-difluorophenyl acetonitrile, benzeneacetonitrile, 2,5-difluoro, 2,5-difluoro-phenyl-acetonitrile, 2-2,5-difluorophenyl ethanenitrile, 2,5-difluorophenylacetonitirle, pubchem4097, 2,5-difluorobenzylcyanide
PubChem CID 592780
IUPAC Name 2-(2,5-difluorophenyl)acetonitrile
SMILES FC1=CC=C(F)C(CC#N)=C1

Specifications

Color Yellow
Boiling Point 90°C
Quantity 5 g
UN Number 3276
Formula Weight 153.13
Percent Purity ≥98.0% (GC)
Physical Form Liquid
Chemical Name or Material 2,5-Difluorobenzyl Cyanide
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.