Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

2,5-Difluoronitrobenzene 98.0+%, TCI America™
SDP

Catalog No. D170025G Shop All Tokyo Chemical Industry (TCI) Products
Change view
Click to view available options
Quantity:
25 g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
D170025G 25 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. D170025G Supplier TCI America Supplier No. D170025G
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

Chemical Identifiers

CAS 364-74-9
Molecular Formula C6H3F2NO2
Molecular Weight (g/mol) 159.09
MDL Number MFCD00007054
InChI Key XNJAYQHWXYJBBD-UHFFFAOYSA-N
Synonym 2,5-difluoronitrobenzene, benzene, 1,4-difluoro-2-nitro, 1,4-difluoro-2-nitro-benzene, pubchem2289, 2,5 difluoronitrobenzene, 2.5-difluoronitrobenzene, acmc-1cjb9, 2,5-difluoro-nitrobenzene, 2,5-difluoronitro-benzene
PubChem CID 67767
IUPAC Name 1,4-difluoro-2-nitrobenzene
SMILES [O-][N+](=O)C1=CC(F)=CC=C1F

Specifications

Boiling Point 104°C
Quantity 25 g
Formula Weight 159.09
Percent Purity ≥98.0% (GC)
Physical Form Liquid
Chemical Name or Material 2,5-Difluoronitrobenzene
RTECSNumber CZ5715000
TSCA No
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.