Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

2,5-Difluorophenylhydrazine, 97%, Thermo Scientific™

Catalog No. AAB2125906
Change view
Click to view available options
Quantity:
5 g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AAB2125906 5 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. AAB2125906 Supplier Thermo Scientific Chemicals Supplier No. B2125906

This Thermo Scientific brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 97108-50-4
Molecular Formula C6H6F2N2
Molecular Weight (g/mol) 144.125
MDL Number MFCD00013384
InChI Key JNBRDOQHPXUXLY-UHFFFAOYSA-N
Synonym 2,5-difluorophenyl hydrazine, 1,4-difluoro-2-hydrazinobenzene, 2,5-difluoro-phenyl-hydrazine, hydrazine, 2,5-difluorophenyl, 2,5-difluorphenylhydrazine hydrochloride, pubchem2467, acmc-20a3z5, 2,5-difluoro-phenylhydrazine, ksc493g8n, 2,5-difluorophenylhydrazine
PubChem CID 588957
IUPAC Name (2,5-difluorophenyl)hydrazine
SMILES C1=CC(=C(C=C1F)NN)F

Specifications

Melting Point 73°C to 76°C
Quantity 5 g
Formula Weight 144.13
Percent Purity 97%
Chemical Name or Material 2,5-Difluorophenylhydrazine
Hazard Category H302+H312+H332-H315-H319-H335
Hazard Statement GHS H Statement
H302-H315-H319-H335
Harmful if swallowed.
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
Precautionary Statement P261-P264b-P270-P271-P280-P301+P312-P302+P352-P304+P340-P305+P351+P338-P312-P330-P332+P313-P362-P501c
DOTInformation Hazard Class: 6.1; Packaging Group: III
TSCA No
Recommended Storage Ambient temperatures
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.