Learn More
Medchemexpress LLC 2,5-Dihydroxyacetophenone | 490-78-8 | 152.15 | 1 ML
SDP
Catalog No. 5000447857
Encompass_Preferred
2,5-Dihydroxyacetophenone, isolated from *Rehmannia glutinosa*, acts as an inhibitor of inflammatory mediators. It blocks the ERK1/2 and NF-κB signaling pathways.
  • Derived from natural source (*Rehmannia glutinosa*).
  • Targets multiple inflammatory pathways (ERK1/2 and NF-κB).
  • Effective in suppressing inflammatory responses in activated macrophages.
Catalog No. 50-004-47857 Supplier Medchemexpress LLC Supplier No. HYW00117410MM/1ML
2,5-Dihydroxyacetophenone, isolated from *Rehmannia glutinosa*, acts as an inhibitor of inflammatory mediators. It blocks the ERK1/2 and NF-κB signaling pathways.
  • Derived from natural source (*Rehmannia glutinosa*).
  • Targets multiple inflammatory pathways (ERK1/2 and NF-κB).
  • Effective in suppressing inflammatory responses in activated macrophages.