Learn More
Cayman Chemical 2,5-Dihydroxybenzaldehyde, Purity: ≥98%, CAS: 1194-98-5, 10 g
SDP
Catalog No. 502978989
Encompass_Preferred
A building block; has been used in the synthesis of 2,4-dimethylbenzoylhydrazones with antileishmanial and antioxidant activities
Catalog No. 50-297-8989 Supplier Cayman Chemical Supplier No. 3026810 G
A building block; has been used in the synthesis of 2,4-dimethylbenzoylhydrazones with antileishmanial and antioxidant activities