Learn More
Medchemexpress LLC HY-W001179 100mg Medchemexpress, 2,5-Dihydroxybenzoic acid CAS:490-79-9 Purity:>98%
SDP
Catalog No. 501934466
Encompass_Preferred
Medchemexpress, HY-W001179 100mg 2,5-Dihydroxybenzoic acid CAS:490-79-9 2,5-Dihydroxybenzoic acid is a derivative of benzoic and a powerful inhibitor of fibroblast growth factors . Purity:>98% Medchemexpress has over 10000 novel life-science reagents, reference compounds, APIs and natural compounds for laboratory and scientific use. Other quantity can also be offered.
Catalog No. 50-193-4466 Supplier Medchemexpress LLC Supplier No. HYW001179100MG
Medchemexpress, HY-W001179 100mg 2,5-Dihydroxybenzoic acid CAS:490-79-9 2,5-Dihydroxybenzoic acid is a derivative of benzoic and a powerful inhibitor of fibroblast growth factors . Purity:>98% Medchemexpress has over 10000 novel life-science reagents, reference compounds, APIs and natural compounds for laboratory and scientific use. Other quantity can also be offered.