Learn More
Sigma Aldrich Fine Chemicals Biosciences 2,5-Dihydroxybenzoic acid matrix substance for MALDI-MS, >=99.5% (HPLC), Ultra pure | 490-79-9 | MFCD00002460 | 10X10MG
Catalog No. NC1608430
Encompass
2,5-Dihydroxybenzoic acid matrix substance for MALDI-MS, >=99.5% (HPLC), Ultra pure | Purity: >=99.5% (HPLC) | Mol Wt: 154.12 | 490-79-9 | MFCD00002460 | 10X10MG
Catalog No. NC1608430 Supplier Sigma Aldrich Fine Chemicals Biosciences Supplier No. 3931910X10MGF
2,5-Dihydroxybenzoic acid matrix substance for MALDI-MS, >=99.5% (HPLC), Ultra pure | Purity: >=99.5% (HPLC) | Mol Wt: 154.12 | 490-79-9 | MFCD00002460 | 10X10MG