Learn More
Medchemexpress LLC 2,5-Dihydroxybenzoic acid | 490-79-9 | 154.12 | 1 ML
SDP
Catalog No. 5000446092
Encompass_Preferred
2,5-Dihydroxybenzoic acid is an analytical standard and a derivative of benzoic acid, intended for research and analytical applications. This compound acts as a powerful inhibitor of fibroblast growth factors, a reactive oxygen species (ROS) scavenger, and an antioxidant. It has demonstrated anti-proliferative activity in human breast cancer cells and inhibits tumor growth in mice with advanced gastric cancer.
  • Analytical standard for research and analytical applications
  • Derivative of benzoic acid
  • Powerful inhibitor of fibroblast growth factors
  • Reactive oxygen species (ROS) scavenger
  • Antioxidant, protecting cells against oxidative damage
  • Exhibits anti-proliferative activity in human breast cancer cells
  • Inhibits tumor growth in mice with advanced gastric cancer
Catalog No. 50-004-46092 Supplier Medchemexpress LLC Supplier No. HYW00117910MM/1ML
2,5-Dihydroxybenzoic acid is an analytical standard and a derivative of benzoic acid, intended for research and analytical applications. This compound acts as a powerful inhibitor of fibroblast growth factors, a reactive oxygen species (ROS) scavenger, and an antioxidant. It has demonstrated anti-proliferative activity in human breast cancer cells and inhibits tumor growth in mice with advanced gastric cancer.
  • Analytical standard for research and analytical applications
  • Derivative of benzoic acid
  • Powerful inhibitor of fibroblast growth factors
  • Reactive oxygen species (ROS) scavenger
  • Antioxidant, protecting cells against oxidative damage
  • Exhibits anti-proliferative activity in human breast cancer cells
  • Inhibits tumor growth in mice with advanced gastric cancer