Learn More
Medchemexpress LLC 2,5-Dihydroxybenzoic acid | 490-79-9 | 154.12 | 500 MG
SDP
Catalog No. 5000447007
Encompass_Preferred
2,5-Dihydroxybenzoic acid, also known as Gentisic acid, is a benzoic acid derivative and a potent inhibitor of fibroblast growth factors. This compound is also a byproduct of aspirin metabolism and found in traditional remedies. It serves as a valuable lead for developing new fibroblast growth factor inhibitors, which may be useful in treating cancer and angiogenesis-dependent diseases.
  • Derivative of benzoic acid
  • Potent inhibitor of fibroblast growth factors
  • Metabolic breakdown product of aspirin
  • Component of traditional liquors and herbal remedies
  • Used as a lead for new fibroblast growth factor inhibitors
  • May be of interest to treat cancer and angiogenesis-dependent diseases
  • Solid appearance
Catalog No. 50-004-47007 Supplier Medchemexpress LLC Supplier No. HYW001179500MG
2,5-Dihydroxybenzoic acid, also known as Gentisic acid, is a benzoic acid derivative and a potent inhibitor of fibroblast growth factors. This compound is also a byproduct of aspirin metabolism and found in traditional remedies. It serves as a valuable lead for developing new fibroblast growth factor inhibitors, which may be useful in treating cancer and angiogenesis-dependent diseases.
  • Derivative of benzoic acid
  • Potent inhibitor of fibroblast growth factors
  • Metabolic breakdown product of aspirin
  • Component of traditional liquors and herbal remedies
  • Used as a lead for new fibroblast growth factor inhibitors
  • May be of interest to treat cancer and angiogenesis-dependent diseases
  • Solid appearance