Learn More
Medchemexpress LLC 2,5-Dihydroxybenzoic acid | 490-79-9 | MFCD00002476 | 154.12 | 1 G
SDP
Catalog No. 5000446091
Encompass_Preferred
2,5-Dihydroxybenzoic acid, also known as Gentisic acid, is a benzoic acid derivative and a potent inhibitor of fibroblast growth factors. It is recognized as a minor metabolite of aspirin and a constituent in various traditional liquors and herbal remedies. This compound serves as a critical lead in the discovery of new chemical classes of fibroblast growth factor inhibitors.
  • Derivative of benzoic acid
  • Potent inhibitor of fibroblast growth factors
  • Minor product of aspirin's metabolic breakdown
  • Component of traditional liquors and herbal remedies
  • Used as a lead to identify new fibroblast growth factor inhibitors
  • May interfere with assembly of signaling complexes for fibroblast growth factor-driven mitogenesis
  • Potential interest for treating cancer and angiogenesis-dependent diseases
Catalog No. 50-004-46091 Supplier Medchemexpress LLC Supplier No. HYW0011791G
2,5-Dihydroxybenzoic acid, also known as Gentisic acid, is a benzoic acid derivative and a potent inhibitor of fibroblast growth factors. It is recognized as a minor metabolite of aspirin and a constituent in various traditional liquors and herbal remedies. This compound serves as a critical lead in the discovery of new chemical classes of fibroblast growth factor inhibitors.
  • Derivative of benzoic acid
  • Potent inhibitor of fibroblast growth factors
  • Minor product of aspirin's metabolic breakdown
  • Component of traditional liquors and herbal remedies
  • Used as a lead to identify new fibroblast growth factor inhibitors
  • May interfere with assembly of signaling complexes for fibroblast growth factor-driven mitogenesis
  • Potential interest for treating cancer and angiogenesis-dependent diseases