Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Sigma Aldrich 2,5-Dilfuoro-4-Methoxyphenylzinc Bromide Solution

Catalog No. 76612725ML Shop All MilliporeSigma Sigma Organic Chemistry Products
Encompass
Encompass_Preferred
Change view
Click to view available options
Quantity:
1 g
25 mL
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
76-612-725ML 1 g
76-612-750ML 25 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Danger
Catalog No. 76-612-725ML Supplier Sigma Aldrich Supplier No. 76612725ML
May include imposed supplier surcharges.
Add to Cart
Add to Cart
Danger

MilliporeSigma Sigma Organics products encompass a wide range of quality reagents, solvents, catalysts, and building blocks for organic synthesis. From benchtop discovery to process development and scale-up, Sigma Organics solutions are built to meet the needs of synthetic chemists.

Nucleophlic reagent applicable for substitution and addition reactions. Also could be used in cross coupling reactions (Negishi couplings).Organozinc ReagentsProduct of Rieke Metals, Inc. Rieke is a registered trademark of Rieke Metals, Inc.

Specifications

CAS 102690-88-0
Density 1.039 g/mL at 25°C
Molecular Formula C12H22N3O2P
Linear Formula C12H22N3O2P
MDL Number MFCD00797597
Quantity 1 g
Molecular Weight (g/mol) 271.3 g/mol
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.