Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

2,5-Dimethoxy-2,5-dihydrofuran, 97%, mixture of cis and trans

Catalog No. AC115531000
Change view
Click to view available options
Quantity:
100 g
Packaging:
Glass bottle
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity Packaging
AC115531000 100 g Glass bottle
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. AC115531000 Supplier Thermo Scientific Chemicals Supplier No. 115531000
Get Up to $1,800 in Chemicals with Purchase
View all promotions
Add to Cart

missing translation for 'validMainframeMsg'

CAS: 332-77-4 | C6H10O3 | 130.14 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 332-77-4
Molecular Formula C6H10O3
Molecular Weight (g/mol) 130.14
MDL Number MFCD00003220
InChI Key WXFWXFIWDGJRSC-UHFFFAOYNA-N
Synonym 2,5-dihydro-2,5-dimethoxyfuran, furan, 2,5-dihydro-2,5-dimethoxy, 2,5-dihydro-2,5-dimethoxyfuran, cis+trans, 2,5-dihydro-2,5-dimethoxyfuran, mixture of cis and trans, pubchem6933, ksc490q3d, furan,5-dihydro-2,5-dimethoxy, 2,5-dihydro-2,5-dimethoxyfuran,c&t, 2,5-dimethoxy-2,5-dihydrofuran,c&t, 2,5-dihydro-2,5-dimethoxyfuran, cis + trans
PubChem CID 78974
IUPAC Name 2,5-dimethoxy-2,5-dihydrofuran
SMILES COC1OC(OC)C=C1

Specifications

CAS Min % 98.5
CAS Max % 100.0
Density 1.0700g/mL
Boiling Point 160.0°C to 162.0°C
Flash Point 47°C
Infrared Spectrum Authentic
Assay Percent Range 96% min. (GC)
Packaging Glass bottle
Refractive Index 1.4330 to 1.4360
Quantity 100 g
Specific Gravity 1.07
Solubility Information Solubility in water: 368g/L (20°C)
Viscosity 2.1 mPa.s (20°C)
Formula Weight 130.14
Percent Purity 97%
Physical Form Liquid
Chemical Name or Material 2,5-Dimethoxy-2,5-dihydrofuran, 99%
Show More Show Less
Hazard Category GHS Signal Word: Warning
EINECSNumber 206-367-5

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.