Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

2',5'-Dimethoxypropiophenone 98.0+%, TCI America™
SDP

Catalog No. D4817200MG Shop All Tokyo Chemical Industry (TCI) Products
Change view
Click to view available options
Quantity:
200 mg
1 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
D4817200MG 200 mg
D48171G 1 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. D4817200MG Supplier TCI America Supplier No. D4817200MG
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

Chemical Identifiers

CAS 5803-30-5
Molecular Formula C11H14O3
Molecular Weight (g/mol) 194.23
MDL Number MFCD00027013
InChI Key DKVCHMQHUMLKPO-UHFFFAOYSA-N
PubChem CID 79865
IUPAC Name 1-(2,5-dimethoxyphenyl)propan-1-one
SMILES CCC(=O)C1=C(C=CC(=C1)OC)OC

Specifications

Color Yellow
Boiling Point 168°C
Quantity 200 mg
Formula Weight 194.23
Percent Purity ≥98.0% (GC)
Physical Form Liquid
Chemical Name or Material 2′,5′-Dimethoxypropiophenone
RTECSNumber UH0530000
TSCA No
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.