Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

2,5-Dimethoxytetrahydrofuran, 99%, mixture of cis- and trans isomers

Catalog No. p-114355
Change view
Click to view available options
Quantity:
100 g
500 g
Packaging:
Glass bottle
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity Packaging
AC115671000 100 g Glass bottle
AC115675000 500 g Glass bottle
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. AC115671000 Supplier Thermo Scientific Chemicals Supplier No. 115671000
Only null left
Add to Cart

CAS: 696-59-3 | C6H12O3 | 132.16 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 696-59-3
Molecular Formula C6H12O3
Molecular Weight (g/mol) 132.16
MDL Number MFCD00005359
InChI Key GFISDBXSWQMOND-UHFFFAOYSA-N
Synonym 2,5-dimethoxytetrahydrofuran, tetrahydro-2,5-dimethoxyfuran, furan, tetrahydro-2,5-dimethoxy, 2,5-dimethoxytetrahydrofurane, 2,5-dimethoxy-tetrahydrofuran, 2,5-dimethoxytetrahydrofuran, mixture of cis-and trans isomers, dimethoxytetrahydrofuran van, protectol dmt, furan,5-dimethoxy, dmthf
PubChem CID 79098
IUPAC Name 2,5-dimethoxyoxolane
SMILES COC1CCC(O1)OC

Specifications

Melting Point -45°C
Color Colorless to Yellow
CAS Min % 98.5
CAS Max % 100.0
Density 1.0210g/mL
Quantity 100 g
Infrared Spectrum Authentic
Packaging Glass bottle
Physical Form Liquid
Refractive Index 1.4160 to 1.419
Assay Percent Range 99%
Fieser 02,144
Solubility Information Solubility in water: 350g/L (20°C). Other solubilities: miscible with alcohols and most common organic,solvents
Boiling Point 145°C to 148°C
Formula Weight 132.16
Percent Purity 99%
Chemical Name or Material 2, 5-Dimethoxytetrahydrofuran, 99%
Show More Show Less
Hazard Category GHS Signal Word: Danger
Hazard Statement GHS H Statement
Flammable liquid and vapor.
Toxic if inhaled.
Causes serious eye irritation.
Precautionary Statement GHS P Statement
Wear eye protection/face protection.
IF ON SKIN (or hair): Take off immediately all contaminated clothing.
Rinse skin with water/shower.
Keep away from heat/sparks/open flames/hot surfaces.
- No smoking.
IF INH
EINECSNumber 211-797-1
TSCA TSCA

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.