Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Medchemexpress LLC 2,5-dimethyl-1,4-benzoquinone | 137-18-8 | MFCD00041737 | 99.4% | 136.15 g/mol | C8H8O2 | 25 G
SDP

Catalog No. 5000208747
Encompass_Preferred

2,5-dimethyl-1,4-benzoquinone is a small organic quinone used as a research reagent and a jack bean urease inhibitor for laboratory studies, including soil urease activity assays.

  • Purity 99.35% by HPLC
  • Chemical formula C8H8O2
  • Molecular weight 136.15 g/mol
  • Available as solid and as 10 mM solutions in DMSO
  • Intended for research use only; not for human or clinical use

Catalog No. 50-002-08747 Supplier Medchemexpress LLC Supplier No. 25GHYW090317
May include imposed supplier surcharges.
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

2,5-dimethyl-1,4-benzoquinone is a small organic quinone used as a research reagent and a jack bean urease inhibitor for laboratory studies, including soil urease activity assays.

  • Purity 99.35% by HPLC
  • Chemical formula C8H8O2
  • Molecular weight 136.15 g/mol
  • Available as solid and as 10 mM solutions in DMSO
  • Intended for research use only; not for human or clinical use

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.