Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

2,5-Dimethyl-2,4-hexadiene (stabilized with BHT) 96.0+%, TCI America™
SDP

Catalog No. D0735500ML Shop All Tokyo Chemical Industry (TCI) Products
Change view
Click to view available options
Quantity:
25 mL
500 mL
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
D0735500ML 500 mL
D073525ML 25 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. D0735500ML Supplier TCI America Supplier No. D0735500ML
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

Chemical Identifiers

CAS 764-13-6
Molecular Formula C8H14
Molecular Weight (g/mol) 110.20
MDL Number MFCD00008904
InChI Key DZPCYXCBXGQBRN-UHFFFAOYSA-N
Synonym 2,5-dimethyl-2,4-hexadiene, biisobutenyl, biisocrotyl, diisocrotyl, 2,4-hexadiene, 2,5-dimethyl, unii-ee5yri32x6, ee5yri32x6, 2,4-diene, acmc-1bi1p, 2,5dimethyl-2,4-hexadiene
PubChem CID 12992
IUPAC Name 2,5-dimethylhexa-2,4-diene
SMILES CC(C)=CC=C(C)C

Specifications

Color Yellow
Boiling Point 135°C
Quantity 500 mL
UN Number 3295
Formula Weight 110.20
Percent Purity ≥96.0% (GC)
Physical Form Liquid
Chemical Name or Material 2,5-Dimethyl-2,4-hexadiene (stabilized with BHT)
EINECSNumber (2)-0028
TSCA Yes
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.