Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

2,5-Dimethyl-3-furanthiol 97.0+%, TCI America™
SDP

Catalog No. D37645G Shop All Tokyo Chemical Industry (TCI) Products
Change view
Click to view available options
Quantity:
1 g
5 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
D37645G 5 g
D37641G 1 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. D37645G Supplier TCI America Supplier No. D37645G
Add to Cart
Add to Cart

Chemical Identifiers

CAS 55764-23-3
Molecular Formula C6H8OS
Molecular Weight (g/mol) 128.19
MDL Number MFCD00801026
InChI Key DBBHCZMXKBCICL-UHFFFAOYSA-N
Synonym 2,5-dimethyl-3-furanthiol, 3-mercapto-2,5-dimethylfuran, 3-furanthiol, 2,5-dimethyl, unii-wct2259x5l, 2,5-dimethyl-3-mercaptofuran, fema no. 3451, 2,5-dimethyl-3-furylmercaptan, 2,5-dimethyl-3-furan thiol, 2,5-dimethyl-furan-3-thiol, acmc-1amqa
PubChem CID 41569
IUPAC Name 2,5-dimethylfuran-3-thiol
SMILES CC1=CC(S)=C(C)O1

Specifications

Boiling Point 150°C
Quantity 5 g
UN Number 3336
Formula Weight 128.19
Percent Purity ≥97.0% (GC)
Physical Form Liquid
Chemical Name or Material 2,5-Dimethyl-3-furanthiol
RTECSNumber LU6230000
TSCA No
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.