Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

2,5-Dimethyl-3-hexanone 96.0+%, TCI America™
SDP

Catalog No. D14535ML Shop All Tokyo Chemical Industry (TCI) Products
Change view
Click to view available options
Quantity:
5 mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
D14535ML 5 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. D14535ML Supplier TCI America Supplier No. D14535ML

missing translation for 'validMainframeMsg'

Chemical Identifiers

CAS 1888-57-9
Molecular Formula C8H16O
Molecular Weight (g/mol) 128.215
MDL Number MFCD00048317
InChI Key TUIWMHDSXJWXOH-UHFFFAOYSA-N
Synonym Isobutyl Isopropyl Ketone
PubChem CID 15901
IUPAC Name 2,5-dimethylhexan-3-one
SMILES CC(C)CC(=O)C(C)C

Specifications

Color Yellow
Boiling Point 148°C
Quantity 5 mL
UN Number 1224
Formula Weight 128.22
Percent Purity ≥96.0% (GC)
Physical Form Liquid
Chemical Name or Material 2,5-Dimethyl-3-hexanone
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.