Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

2,5-Dimethyl-4-(morpholinomethyl)phenol Hydrochloride Monohydrate 97%, Thermo Scientific™

Catalog No. AC181600250
Change view
Click to view available options
Quantity:
25 g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AC181600250 25 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. AC181600250 Supplier Thermo Scientific Chemicals Supplier No. 181600250

missing translation for 'validMainframeMsg'

This Thermo Scientific brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 33625-43-3
Molecular Formula C13H20ClNO2
Molecular Weight (g/mol) 257.76
MDL Number MFCD00012796
InChI Key RLGUYDSHQWRANT-UHFFFAOYSA-N
Synonym 2,5-dimethyl-4-morpholinomethyl phenol hydrochloride, phenol, 2,5-dimethyl-4-4-morpholinylmethyl-, hydrochloride, phenol, 2,5-dimethyl-4-4-morpholinylmethyl-, hydrochloride 1:1, 2,5-dimethyl-4-morpholin-4-ylmethyl phenol hydrochloride, 2,5-dimethyl-4-morpholin-4-ylmethyl phenol, chloride, 4-morpholinomethyl-2,5-xylenol hydrochloride, acmc-20aoah, 2,5-dimethyl-4-morpholinomethyl phenolhydrochloride, 2,5-dimethyl-4-4-morpholinylmethyl phenol hydrochloride hydrate
PubChem CID 118512
IUPAC Name 2,5-dimethyl-4-[(morpholin-4-yl)methyl]phenol hydrochloride
SMILES Cl.CC1=CC(CN2CCOCC2)=C(C)C=C1O

Specifications

Color White
CAS Min % 96.0
CAS Max % 100.0
Quantity 25 g
Infrared Spectrum Authentic
Physical Form Micro-Crystalline Powder
Formula Weight 275.77
Percent Purity ≥96.0%
Chemical Name or Material 2, 5-Dimethyl-4-(morpholinomethyl)phenol hydrochloride monohydrate, 97%
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.