Learn More
AdipoGen 2,5-Dimethyl-celecoxib (25 mg)
SDP
Supplier:  AdipoGen AGCR13599M025
Encompass_Preferred
Chemical. CAS: 457639-26-8. Formula: C18H16F3N3O2S. MW: 395.4. Close structural analog of the selective cyclooxygenase-2 (COX-2) inhibitor celecoxib, that lacks COX-2 inhibitory function. Anti-proliferative, anti-tumorigenic and anti-angiogenic. Potent apoptosis inducer in many cancer cell lines. Down-regulates the expression of survivin, cyclins, MMPs and inhibits cyclin-dependent kinase activity. Modulates PDK-1, AKT, GSK3beta, p70 S6K, PKA and MAPKAP-K1alpha. Reduces phosphorylation of ERK1/2 but not AKT T-308 or AKT S-473. Increases intracellular free calcium levels and potently triggers the endoplasmatic reticulum stress response (ESR), activating ER stress-associated proteins GRP78/BiP, CHOP/GADD153 and caspase-4.
Catalog No. 50-205-3970