Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

2,5-Dimethylbenzyl alcohol, 97+%

Catalog No. AAL0162403
Change view
Click to view available options
Quantity:
1 g
5 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AAL0162403 1 g
AAL0162406 5 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. AAL0162403 Supplier Thermo Scientific Chemicals Supplier No. L0162403
Add to Cart

CAS: 53957-33-8 | C9H12O | 136.194 g/mol

2,5-Dimethylbenzyl alcohol, is used as a self-indicating standard for the determination of organ lithium reagents

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
2,5-Dimethylbenzyl alcohol, is used as a self-indicating standard for the determination of organ lithium reagents

Solubility
Difficult in mix.

Notes
Store away from strong oxidizing agents. Keep container tightly closed. Store in cool, dry conditions in well sealed containers.
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 53957-33-8
Molecular Formula C9H12O
Molecular Weight (g/mol) 136.194
MDL Number MFCD00004625
InChI Key LEBQTCCCNMTXSF-UHFFFAOYSA-N
PubChem CID 94560
IUPAC Name (2,5-dimethylphenyl)methanol
SMILES CC1=CC(=C(C=C1)C)CO

Specifications

Melting Point 40°C to 42°C
Density 1.011
Boiling Point 232°C to 234°C
Flash Point 107°C (224°F)
Refractive Index 1.536
Quantity 1 g
Beilstein 2243420
Synonym 2,5-dimethylbenzyl alcohol, 2,5-dimethylphenyl methanol, benzenemethanol, 2,5-dimethyl, acmc-1ayay, 2,5-dimethylbenzylalcohol, benzenemethanol,2,5-dimethyl, 2,5-dimethylphenyl methanol #, 2'-oxy-1.2.4-trimethylbenzol, 2,5-dimethyl-hydroxymethylbenzene
Solubility Information Difficult in mix.
Formula Weight 136.19
Percent Purity ≥97%
Chemical Name or Material 2,5-Dimethylbenzyl alcohol
Show More Show Less
EINECSNumber 258-888-2
TSCA No
Recommended Storage Ambient temperatures

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.