Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

2,5-Dimethylfuran-3-carbonyl chloride, 97%, Thermo Scientific™

Catalog No. p-2696817
Change view
Click to view available options
Quantity:
1 g
5 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
CC01102DA 1 g
CC01102DE 5 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. CC01102DA Supplier Thermo Scientific Chemicals Supplier No. CC01102DA
Only null left
Add to Cart

Chemical Identifiers

CAS 50990-93-7
Molecular Formula C7H7ClO2
Molecular Weight (g/mol) 158.58
MDL Number MFCD00221073
InChI Key RWORXZHVOUPMMM-UHFFFAOYSA-N
Synonym 2,5-dimethyl-3-furoyl chloride, 2,5-dimethyl-furan-3-carbonyl chloride, 2,5-dimethyl-3-furancarbonyl chloride, 2,5-dimethylfuran-3-carbonylchloride, pubchem6936, acmc-209krc, 2,5-dimethyl-3-furoylchloride, 2,5 dimethyl-furan-3-carbonyl chloride, 2,5-dimethyl furan-3-carbonyl chloride, 3-furancarbonylchloride, 2,5-dimethyl
PubChem CID 2734804
IUPAC Name 2,5-dimethylfuran-3-carbonyl chloride
SMILES CC1=CC(C(Cl)=O)=C(C)O1

Specifications

CAS Min % 96.5
CAS Max % 100.0
Density 1.19g/mL
Assay Percent Range 97%
Packaging Amber glass bottle
Quantity 1 g
Formula Weight 158.58
Percent Purity 97%
Chemical Name or Material 2, 5-Dimethylfuran-3-carbonyl chloride
Hazard Category GHS Signal Word: Danger
Hazard Statement GHS H Statement:
Harmful if inhaled.
Causes severe skin burns and eye damage.
Harmful if swallowed.
Harmful in contact with skin.
Precautionary Statement GHS P Statement:
Wear protective gloves/protective clothing/eye protection/face protection.
IF SWALLOWED: rinse mouth. Do NOT induce vomiting.
Wear eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
Immediately call a POISON CENTER or doctor/physician.
IF ON SKIN: Wash with plenty of soap and water.
IF SWALLOWED: Call a POISON CENTER or doctor/physician if you feel unwell.
IF INHALED: Remove victim to fresh air and keep at rest in a position comfortable for breathing.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.