Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

2,5-Dimethylnitrobenzene 99.0+%, TCI America™
SDP

Catalog No. D1105500G Shop All Tokyo Chemical Industry (TCI) Products
Change view
Click to view available options
Quantity:
25 g
500 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
D1105500G 500 g
D110525G 25 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. D1105500G Supplier TCI America Supplier No. D1105500G
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

CAS: 89-58-7 | C8H9NO2 | 151.17 g/mol

Chemical Identifiers

CAS 89-58-7
Molecular Formula C8H9NO2
Molecular Weight (g/mol) 151.17
MDL Number MFCD00024284
InChI Key BSFHJMGROOFSRA-UHFFFAOYSA-N
Synonym Nitro-p-xylene
PubChem CID 6974
IUPAC Name 1,4-dimethyl-2-nitrobenzene
SMILES CC1=CC=C(C)C(=C1)[N+]([O-])=O

Specifications

Color Yellow
Quantity 500 g
Physical Form Clear Liquid at 20°C
UN Number 1665
Boiling Point 104°C
Formula Weight 151.17
Percent Purity ≥99.0% (GC)
Grade Not Available
Chemical Name or Material 2,5-Dimethylnitrobenzene
EINECSNumber (3)-0399
RTECSNumber ZE4686600
TSCA Yes
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.