Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

(2,5-Dimethylphenyl)magnesium Bromide (20% in Tetrahydrofuran, ca. 1mol/L), TCI America™
SDP

Catalog No. D3551100G Shop All Tokyo Chemical Industry (TCI) Products
Change view
Click to view available options
Quantity:
100 g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
D3551100G 100 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. D3551100G Supplier TCI America Supplier No. D3551100G

missing translation for 'validMainframeMsg'

Chemical Identifiers

CAS 30897-86-0
Molecular Formula C8H9BrMg
Molecular Weight (g/mol) 209.37
MDL Number MFCD01311495
InChI Key OOJAWTIYZUSKLZ-UHFFFAOYSA-M
Synonym 2,5-dimethylphenylmagnesium bromide, 2,5-xylylmagnesium bromide, 2,5-dimethylphenyl magnesium bromide, p-xylene-2-magnesium bromide, xylkrqfnmxvtlq-uhfffaoysa-m, 2,5-dimethylphenyl magnesiumbromide, 2,5-dimethylphenylmagnesium bromide, 0.5 m in thf, 2,5-dimethylphenylmagnesium bromide 0.5 m in tetrahydrofuran
PubChem CID 10976718
IUPAC Name magnesium(2+) 2,5-dimethylbenzen-1-ide bromide
SMILES [Mg++].[Br-].CC1=CC=C(C)[C-]=C1

Specifications

Boiling Point 65°C
UN Number 2924
Quantity 100 g
Formula Weight 209.37
Chemical Name or Material (2,5-Dimethylphenyl)magnesium Bromide (20% in Tetrahydrofuran, ca. 1mol/L)
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.