Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

2,5-Dimethylphenylacetyl Chloride 98.0+%, TCI America™
SDP

Catalog No. D42615G Shop All Tokyo Chemical Industry (TCI) Products
Change view
Click to view available options
Quantity:
5 g
25 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
D42615G 5 g
D426125G 25 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. D42615G Supplier TCI America Supplier No. D42615G

Chemical Identifiers

CAS 55312-97-5
Molecular Formula C10H11ClO
Molecular Weight (g/mol) 182.647
MDL Number MFCD12024950
InChI Key PZRANTBLOIKHJY-UHFFFAOYSA-N
PubChem CID 22561615
IUPAC Name 2-(2,5-dimethylphenyl)acetyl chloride
SMILES CC1=CC(=C(C=C1)C)CC(=O)Cl

Specifications

Color Yellow
Quantity 5 g
Physical Form Clear Liquid at 20°C
UN Number 3265
Formula Weight 182.65
Percent Purity ≥98.0% (GC,T)
Chemical Name or Material 2,5-Dimethylphenylacetyl Chloride
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.