Learn More
2,5-Dimethylphenylboronic Acid (contains varying amounts of Anhydride), TCI America™
Shop All Tokyo Chemical Industry (TCI) Products

Specifications
Specifications
| CAS | 85199-06-0 |
| Quantity | 25 g |
| Physical Form | Crystal-Powder at 20°C |
| Molecular Formula | C8H11BO2 |
| MDL Number | MFCD01863525 |
| Synonym | 2,5-dimethylphenyl boronic acid, 2,5-dimethylbenzeneboronic acid, 2,5-dimethylphenylboronicacid, 2,5-dimethylphenyl boranediol, p-xylene-2-boronic acid |
| InChI Key | OOMZKLJLVGQZGV-UHFFFAOYSA-N |
| SMILES | B(C1=C(C=CC(=C1)C)C)(O)O |
| IUPAC Name | (2,5-dimethylphenyl)boronic acid |
| Molecular Weight (g/mol) | 149.984 |
| Show More |
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.