Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

2,5-Dimethyltetrahydrofuran, Mixture of cis and trans 99%, Thermo Scientific™

Catalog No. AC116841000
Change view
Click to view available options
Quantity:
100 mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AC116841000 100 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. AC116841000 Supplier Thermo Scientific Chemicals Supplier No. 116841000

missing translation for 'validMainframeMsg'

This Thermo Scientific brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 1003-38-9
Molecular Formula C6H12O
Molecular Weight (g/mol) 100.16
MDL Number MFCD00005369
InChI Key OXMIDRBAFOEOQT-UHFFFAOYNA-N
Synonym 2,5-dimethyltetrahydrofuran, furan, tetrahydro-2,5-dimethyl, tetrahydro-2,5-dimethylfuran, 2,5-dimethyltetrahydrofuran, mixture of cis and trans, cis-tetrahydro-2,5-dimethylfuran, acmc-1c5fn, 2,5-dimethyl-tetrahydrofuran, 2,5-dimethyl-tetrahydro-furan, 2,5-dimethyltetrahydrofuran,c&t, 2,5-dimethyltetrahydrofuran,mixtureofcisandtrans
PubChem CID 13855
IUPAC Name 2,5-dimethyloxolane
SMILES CC1CCC(C)O1

Specifications

CAS Min % 98.5
CAS Max % 100.0
Density 0.83
Quantity 100 mL
Beilstein 17, 14
Formula Weight 100.16
Chemical Name or Material 2, 5-Dimethyltetrahydrofuran, mixture of cis and trans, 99%
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.