Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Medchemexpress LLC 2,5-dioxopyrrolidin-1-yl 1-(4-(6-methyl-1,2,4,5-tetrazin-3-yl)phenyl)-PEG4-SS-NHS ester | 95.0% | 737.84 | C31H43N7O10S2 | 10 MG
SDP

Catalog No. 5000202580
Encompass_Preferred

Methyltetrazine-PEG4-SS-NHS ester is a cleavable PEG4-based methyltetrazine-functionalized N-hydroxysuccinimide ester for bioconjugation and click-chemistry applications. It provides a tetrazine reactive handle for inverse electron-demand Diels-Alder labeling, and contains a disulfide cleavable spacer and PEG4 linker to improve solubility and enable reductive release.

  • Purity 95.0% (NMR).
  • Pink to red oil appearance.
  • Molecular weight 737.84.
  • Chemical formula C31H43N7O10S2.
  • Contains a disulfide cleavable linker and a PEG4 spacer.
  • Provides a tetrazine handle for IEDDA bioconjugation.
  • Storage: pure form at -20°C (stable for 3 years); in solvent -80°C for 6 months, -20°C for 1 month.
  • Tested by 1H NMR; for research use only.

Catalog No. 50-002-02580 Supplier Medchemexpress LLC Supplier No. HY13346610MG
May include imposed supplier surcharges.
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

Methyltetrazine-PEG4-SS-NHS ester is a cleavable PEG4-based methyltetrazine-functionalized N-hydroxysuccinimide ester for bioconjugation and click-chemistry applications. It provides a tetrazine reactive handle for inverse electron-demand Diels-Alder labeling, and contains a disulfide cleavable spacer and PEG4 linker to improve solubility and enable reductive release.

  • Purity 95.0% (NMR).
  • Pink to red oil appearance.
  • Molecular weight 737.84.
  • Chemical formula C31H43N7O10S2.
  • Contains a disulfide cleavable linker and a PEG4 spacer.
  • Provides a tetrazine handle for IEDDA bioconjugation.
  • Storage: pure form at -20°C (stable for 3 years); in solvent -80°C for 6 months, -20°C for 1 month.
  • Tested by 1H NMR; for research use only.

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.