Learn More
Medchemexpress LLC 2,5-dioxopyrrolidin-1-yl 1-(bicyclo[6.1.0]non-4-yn-9-yl) pentaoxa-4-azanonadecan-oate | 00-00-0 | 95.0% | C26H38N2O10 | 10MG
SDP
Catalog No. 5000259060
Encompass_Preferred
BCN-PEG4-NHS ester is a polyethylene glycol-based bifunctional linker that pairs a bicyclononyne moiety for copper-free click conjugation with an N-hydroxysuccinimide activated ester for amine coupling. Supplied as a colorless to light yellow liquid with high reported purity, it is designed for bioconjugation, linker assembly, and synthesis workflows that require efficient, selective coupling and good solubility.
  • enables copper-free click conjugation via strain-promoted alkyne-azide cycloaddition.
  • provides an NHS-activated ester for rapid amine coupling.
  • peg4 spacer enhances solubility and reduces steric hindrance.
  • high reported purity suitable for synthetic and bioconjugation use.
  • stable under recommended frozen storage for extended shelf life.
  • available in small pack sizes for method development and optimization.
Catalog No. 50-002-59060 Supplier Medchemexpress LLC Supplier No. HY13343910MG
BCN-PEG4-NHS ester is a polyethylene glycol-based bifunctional linker that pairs a bicyclononyne moiety for copper-free click conjugation with an N-hydroxysuccinimide activated ester for amine coupling. Supplied as a colorless to light yellow liquid with high reported purity, it is designed for bioconjugation, linker assembly, and synthesis workflows that require efficient, selective coupling and good solubility.
  • enables copper-free click conjugation via strain-promoted alkyne-azide cycloaddition.
  • provides an NHS-activated ester for rapid amine coupling.
  • peg4 spacer enhances solubility and reduces steric hindrance.
  • high reported purity suitable for synthetic and bioconjugation use.
  • stable under recommended frozen storage for extended shelf life.
  • available in small pack sizes for method development and optimization.