Learn More
Medchemexpress LLC 2,5-dioxopyrrolidin-1-yl 11-mercaptoundecanoate | 240125-70-6 | MFCD18642190 | 98.0% | 315.4 g/mol | C15H25NO4S | 250 MG
SDP
Catalog No. 5000281342
Encompass_Preferred
2,5-Dioxopyrrolidin-1-yl 11-mercaptoundecanoate is a bifunctional thiol-NHS ester linker designed for conjugation and surface modification. It combines a reactive N-hydroxysuccinimide (NHS) ester for amide bond formation with a free thiol for attachment to thiol-reactive groups or metal surfaces, enabling reliable formation of stable linkages in bioconjugation workflows.
  • Provides an NHS ester for efficient coupling to primary amines.
  • Provides a free thiol for attachment to maleimide, vinylsulfone, or metal surfaces.
  • Suitable for bioconjugation, surface modification, and linker synthesis.
  • Molecular weight about 315.4 g/mol and formula C15H25NO4S.
  • Available as a powder with recommended storage at -20°C for long-term stability.
  • High purity (≥98.0% by NMR) for consistent coupling performance.
Catalog No. 50-002-81342 Supplier Medchemexpress LLC Supplier No. HYW405064250MG
2,5-Dioxopyrrolidin-1-yl 11-mercaptoundecanoate is a bifunctional thiol-NHS ester linker designed for conjugation and surface modification. It combines a reactive N-hydroxysuccinimide (NHS) ester for amide bond formation with a free thiol for attachment to thiol-reactive groups or metal surfaces, enabling reliable formation of stable linkages in bioconjugation workflows.
  • Provides an NHS ester for efficient coupling to primary amines.
  • Provides a free thiol for attachment to maleimide, vinylsulfone, or metal surfaces.
  • Suitable for bioconjugation, surface modification, and linker synthesis.
  • Molecular weight about 315.4 g/mol and formula C15H25NO4S.
  • Available as a powder with recommended storage at -20°C for long-term stability.
  • High purity (≥98.0% by NMR) for consistent coupling performance.