Learn More
Medchemexpress LLC 2,5-dioxopyrrolidin-1-yl 2-(prop-2-yn-1-yloxy)acetate | 1858242-47-3 | MFCD29067173 | >98.0% | 211.17 g·mol⁻¹ | C9H9NO5 | 250 MG
SDP
Catalog No. 5000281741
Encompass_Preferred
2,5-Dioxopyrrolidin-1-yl 2-(prop-2-yn-1-yloxy)acetate is an N-hydroxysuccinimide (NHS) ester bearing a propargyl (alkyne) handle. It is used to introduce terminal alkyne functionality onto amine-containing molecules via NHS-ester coupling for downstream click chemistry and bioconjugation workflows. Reported purity is >98% and common pack sizes include 250 mg.
  • Provides a propargyl handle for copper-catalyzed azide-alkyne cycloaddition (click chemistry).
  • Reacts readily with primary amines through NHS-ester-mediated amide bond formation.
  • High reported purity supports reliable conjugation and analytical reproducibility.
  • Available in small pack sizes suitable for preparative and research use.
  • Stable when stored under recommended conditions for reagent-grade NHS esters.
Catalog No. 50-002-81741 Supplier Medchemexpress LLC Supplier No. HYW451460250MG
2,5-Dioxopyrrolidin-1-yl 2-(prop-2-yn-1-yloxy)acetate is an N-hydroxysuccinimide (NHS) ester bearing a propargyl (alkyne) handle. It is used to introduce terminal alkyne functionality onto amine-containing molecules via NHS-ester coupling for downstream click chemistry and bioconjugation workflows. Reported purity is >98% and common pack sizes include 250 mg.
  • Provides a propargyl handle for copper-catalyzed azide-alkyne cycloaddition (click chemistry).
  • Reacts readily with primary amines through NHS-ester-mediated amide bond formation.
  • High reported purity supports reliable conjugation and analytical reproducibility.
  • Available in small pack sizes suitable for preparative and research use.
  • Stable when stored under recommended conditions for reagent-grade NHS esters.