Learn More
Medchemexpress LLC 2,5-dioxopyrrolidin-1-yl 3-((2-(((((1R,8S,9S)-bicyclo[6.1.0]non-4-yn-9-yl)methoxy)carbonyl)amino)... | 95.0% | 454.56 g/mol | C20H26N2O6S2 | 10 MG
SDP
Catalog No. 5000203370
Encompass_Preferred
BCN-SS-NHS is a cleavable linker reagent containing an NHS ester and a strained bicyclo[6.1.0]nonyne (BCN) moiety. It is used for strain-promoted alkyne-azide cycloaddition (SPAAC) click chemistry and to introduce a cleavable disulfide-containing linker arm in bioconjugation and antibody-drug conjugate synthesis.
  • Used for SPAAC click chemistry
  • Contains an NHS ester for efficient amine coupling
  • Includes a disulfide-cleavable linker for intracellular release
  • Suitable for antibody-drug conjugate and bioconjugation workflows
  • Available in small-scale research quantities
Catalog No. 50-002-03370 Supplier Medchemexpress LLC Supplier No. HY13597310MG
BCN-SS-NHS is a cleavable linker reagent containing an NHS ester and a strained bicyclo[6.1.0]nonyne (BCN) moiety. It is used for strain-promoted alkyne-azide cycloaddition (SPAAC) click chemistry and to introduce a cleavable disulfide-containing linker arm in bioconjugation and antibody-drug conjugate synthesis.
  • Used for SPAAC click chemistry
  • Contains an NHS ester for efficient amine coupling
  • Includes a disulfide-cleavable linker for intracellular release
  • Suitable for antibody-drug conjugate and bioconjugation workflows
  • Available in small-scale research quantities