Learn More
Medchemexpress LLC (2,5-dioxopyrrolidin-1-yl) 3-[2-[2-[2-[2-[2-[2-[2-[2-[2-[2-[2-[2-[2-propanoate | 2395839-96-8 | MFCD11041155 | 99.9% | C54H103NO28 | 10MG
SDP
Catalog No. 5000261875
Encompass_Preferred
m-PEG24-NHS ester is a methyl-capped polyethylene glycol (PEG) linker bearing an N-hydroxysuccinimide (NHS) activated ester. It is designed for coupling to primary amines and is commonly used as a hydrophilic spacer in the synthesis of PROTACs, bioconjugates, and other bifunctional molecules where controlled spacing and solubility are required.
  • Methyl-capped PEG24 spacer with terminal NHS ester for efficient amine coupling.
  • High purity suitable for synthetic and bioconjugation workflows.
  • Hydrophilic and flexible linker improves solubility and reduces steric hindrance.
  • Compatible with standard NHS-ester coupling conditions.
  • Available in laboratory-scale quantities for research and development use.
Catalog No. 50-002-61875 Supplier Medchemexpress LLC Supplier No. HY13083010MG
m-PEG24-NHS ester is a methyl-capped polyethylene glycol (PEG) linker bearing an N-hydroxysuccinimide (NHS) activated ester. It is designed for coupling to primary amines and is commonly used as a hydrophilic spacer in the synthesis of PROTACs, bioconjugates, and other bifunctional molecules where controlled spacing and solubility are required.
  • Methyl-capped PEG24 spacer with terminal NHS ester for efficient amine coupling.
  • High purity suitable for synthetic and bioconjugation workflows.
  • Hydrophilic and flexible linker improves solubility and reduces steric hindrance.
  • Compatible with standard NHS-ester coupling conditions.
  • Available in laboratory-scale quantities for research and development use.