Learn More
Medchemexpress LLC 2,5-dioxopyrrolidin-1-yl acrylate | 38862-24-7 | MFCD00078261 | 99.6% | 169.13 g/mol | C7H7NO4 | 250 MG
SDP
Catalog No. 5000209187
Encompass_Preferred
2,5-dioxopyrrolidin-1-yl acrylate (N-succinimidyl acrylate) is an amine-reactive N-hydroxysuccinimide (NHS) ester that introduces acrylate functionality onto primary amines for bioconjugation and polymerization applications. It is commonly used to modify lysine residues on proteins and to install polymerizable acrylate groups for downstream coupling or surface modification.
  • Reacts with primary amines to form amide linkages.
  • Introduces acrylate groups for polymerization or click-type chemistry.
  • High reported purity suitable for research use.
  • Soluble in common organic solvents and DMSO for convenient handling.
  • Useful for protein conjugation, crosslinking, and surface functionalization.
Catalog No. 50-002-09187 Supplier Medchemexpress LLC Supplier No. 250MGHYW043748
2,5-dioxopyrrolidin-1-yl acrylate (N-succinimidyl acrylate) is an amine-reactive N-hydroxysuccinimide (NHS) ester that introduces acrylate functionality onto primary amines for bioconjugation and polymerization applications. It is commonly used to modify lysine residues on proteins and to install polymerizable acrylate groups for downstream coupling or surface modification.
  • Reacts with primary amines to form amide linkages.
  • Introduces acrylate groups for polymerization or click-type chemistry.
  • High reported purity suitable for research use.
  • Soluble in common organic solvents and DMSO for convenient handling.
  • Useful for protein conjugation, crosslinking, and surface functionalization.