Learn More
Medchemexpress LLC 2,5-dioxopyrrolidin-1-yl hept-6-ynoate | 917222-23-2 | MFCD29041721 | 98.0% | 223.23 g/mol | C11H13NO4 | 250 MG
SDP
Catalog No. 5000281477
Encompass_Preferred
2,5-Dioxopyrrolidin-1-yl hept-6-ynoate is an amine-reactive N-hydroxysuccinimide (NHS) ester bearing a terminal alkyne. It enables selective derivatization of primary amines on peptides, proteins, antibodies, and amine-coated surfaces and facilitates copper-catalyzed azide-alkyne cycloaddition (Click Chemistry) for bioconjugation workflows.
  • Amine-reactive N-hydroxysuccinimide (NHS) ester for selective labeling of primary amines.
  • Terminal alkyne enables copper-catalyzed azide-alkyne cycloaddition (Click Chemistry).
  • Suitable for peptides, proteins, antibodies, and amine-coated surfaces.
  • Available in small laboratory pack sizes for method development and optimization.
  • Recommended storage under cool, inert conditions to maintain stability.
Catalog No. 50-002-81477 Supplier Medchemexpress LLC Supplier No. HYW451405250MG
2,5-Dioxopyrrolidin-1-yl hept-6-ynoate is an amine-reactive N-hydroxysuccinimide (NHS) ester bearing a terminal alkyne. It enables selective derivatization of primary amines on peptides, proteins, antibodies, and amine-coated surfaces and facilitates copper-catalyzed azide-alkyne cycloaddition (Click Chemistry) for bioconjugation workflows.
  • Amine-reactive N-hydroxysuccinimide (NHS) ester for selective labeling of primary amines.
  • Terminal alkyne enables copper-catalyzed azide-alkyne cycloaddition (Click Chemistry).
  • Suitable for peptides, proteins, antibodies, and amine-coated surfaces.
  • Available in small laboratory pack sizes for method development and optimization.
  • Recommended storage under cool, inert conditions to maintain stability.