Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Medchemexpress LLC (2,5-dioxopyrrolidin-1-yl) hex-5-ynoate | 906564-59-8 | MFCD28898858 | 97.0% | 209.20 g/mol | C10H11NO4 | 50 MG
SDP

Catalog No. 5000204048
Encompass_Preferred

Propargyl-C2-NHS ester is an alkyne-containing N-hydroxysuccinimide (NHS) ester used for bioconjugation. It serves as a non-cleavable linker and a click-chemistry reagent enabling copper-catalyzed azide-alkyne cycloaddition (CuAAC) with azide-bearing molecules.

  • Contains a propargyl (alkyne) group for CuAAC reactions.
  • Designed for antibody conjugation and general bioconjugation.
  • Molecular formula C10H11NO4; molecular weight 209.20 g/mol.
  • High purity: 97.0%.
  • Stored at -20°C in pure form; available in small pack sizes for research use.

Catalog No. 50-002-04048 Supplier Medchemexpress LLC Supplier No. HY12651250MG
May include imposed supplier surcharges.
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

Propargyl-C2-NHS ester is an alkyne-containing N-hydroxysuccinimide (NHS) ester used for bioconjugation. It serves as a non-cleavable linker and a click-chemistry reagent enabling copper-catalyzed azide-alkyne cycloaddition (CuAAC) with azide-bearing molecules.

  • Contains a propargyl (alkyne) group for CuAAC reactions.
  • Designed for antibody conjugation and general bioconjugation.
  • Molecular formula C10H11NO4; molecular weight 209.20 g/mol.
  • High purity: 97.0%.
  • Stored at -20°C in pure form; available in small pack sizes for research use.

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.