Learn More
Medchemexpress LLC 2,5-dioxopyrrolidin-1-yl m-PEG24 carboxylate | 2395839-96-8 | MFCD11041155 | 99.7% | 1214.39 g·mol⁻¹ | C54H103NO28 | 5 MG
SDP
Catalog No. 5000262157
Encompass_Preferred
m-PEG24-NHS ester is a methoxy-terminated polyethylene glycol (PEG) linker functionalized with an N-hydroxysuccinimide (NHS) ester. The NHS group reacts with primary amines to form stable amide bonds, enabling bioconjugation and linker installation in medicinal chemistry workflows such as PROTAC assembly. The reagent is provided with high purity for research applications.
  • Methoxy-terminated PEG24 linker with NHS ester reactivity.
  • Reacts with primary amines to form stable amide bonds.
  • Compatible with PROTAC synthesis and general bioconjugation.
  • High purity suitable for research use.
  • Molecular weight approximately 1214.39 g·mol⁻¹.
Catalog No. 50-002-62157 Supplier Medchemexpress LLC Supplier No. HY1308305MG
m-PEG24-NHS ester is a methoxy-terminated polyethylene glycol (PEG) linker functionalized with an N-hydroxysuccinimide (NHS) ester. The NHS group reacts with primary amines to form stable amide bonds, enabling bioconjugation and linker installation in medicinal chemistry workflows such as PROTAC assembly. The reagent is provided with high purity for research applications.
  • Methoxy-terminated PEG24 linker with NHS ester reactivity.
  • Reacts with primary amines to form stable amide bonds.
  • Compatible with PROTAC synthesis and general bioconjugation.
  • High purity suitable for research use.
  • Molecular weight approximately 1214.39 g·mol⁻¹.